Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nipal2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145469.5 | NP_663444.2 | 2 - 3 | 213 | 81 | |
GenBank mRNA | AK033975.1 | - | 2 - 3 | 214 | 81 | |
AK038556.1 | - | 2 - 3 | 354 | 81 | ||
AK079085.1 | - | 2 - 3 | 646 | 81 | ||
AK132530.1 | - | 2 - 3 | 241 | 81 | ||
BC016107.1 | - | 2 - 3 | 236 | 4921508 | 81 | |
BC030399.1 | - | 2 - 3 | 730 | 4166825 | 81 | |
BC058207.1 | - | 2 - 3 | 379 | 5361417 | 81 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |