Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ehmt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001286573.1 | NP_001273502.1 | 5 - 6 | 860 | 56 | |
NM_001286575.1 | NP_001273504.1 | 6 - 7 | 696 | 56 | ||
NM_145830.2 | NP_665829.1 | 5 - 6 | 860 | 56 | ||
NM_147151.2 | NP_671493.1 | 6 - 7 | 696 | 56 | ||
GenBank mRNA | AB077209.1 | - | 6 - 7 | 689 | 56 | |
AB077210.1 | - | 5 - 6 | 860 | 56 | ||
AK154246.1 | - | 5 - 6 | 883 | 56 | ||
BC058357.1 | - | 6 - 7 | 694 | 56 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |