Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc44a4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_023557.3 | NP_076046.1 | 16 - 17 | 1698 | 126 | |
GenBank mRNA | AK008864.1 | - | 5 - 6 | 660 | 126 | |
AK145621.1 | - | 16 - 17 | 1690 | 126 | ||
AK166620.1 | - | 13 - 14 | 1600 | 126 | ||
BC010808.1 | - | 16 - 17 | 1696 | 4240658 | 126 | |
BC052652.1 | - | 16 - 17 | 1696 | 30034507 | 126 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |