Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013484.2 | NP_038512.2 | 1 - 2 | 148 | 80 | |
GenBank mRNA | AK146812.1 | - | 1 - 2 | 149 | 80 | |
AK216863.1 | - | 195 | 80 | |||
BC011086.1 | - | 1 - 2 | 111 | 3992881 | 80 | |
M57891.1 | - | 1 - 2 | 95 | 80 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protease Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |