Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vars | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011690.3 | NP_035820.3 | 12 - 13 | 1927 | 82 | |
GenBank mRNA | AF087680.1 | - | 12 - 13 | 1690 | 82 | |
AK019157.1 | - | 4 - 5 | 81 | |||
AK154823.1 | - | 12 - 13 | 1742 | 82 | ||
AK157814.1 | - | 12 - 13 | 1785 | 82 | ||
AK167842.1 | - | 4 - 5 | 465 | 82 | ||
BC053703.1 | - | 12 - 13 | 1710 | 30055190 | 82 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |