Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc17a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_144836.2 | NP_659085.1 | 8 - 9 | 1342 | 70 | |
GenBank mRNA | AK040717.1 | - | 8 - 9 | 1343 | 70 | |
BC018306.1 | - | 7 - 8 | 980 | 4193755 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Transporter Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |