Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prkag2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001170555.1 | NP_001164026.1 | 7 - 8 | 655 | 132 | |
NM_001170556.1 | NP_001164027.1 | 7 - 8 | 652 | 132 | ||
NM_145401.2 | NP_663376.2 | 11 - 12 | 1734 | 132 | ||
GenBank mRNA | AK032238.1 | - | 9 - 10 | 1246 | 132 | |
AK034765.1 | - | 5 - 6 | 320 | 132 | ||
AK154296.1 | - | 11 - 12 | 1735 | 132 | ||
AK165834.1 | - | 7 - 8 | 594 | 132 | ||
AY348864.1 | - | 7 - 8 | 585 | 132 | ||
BC015283.1 | - | 11 - 12 | 1522 | 4238045 | 132 | |
BQ572978.1 | - | 7 - 8 | 585 | 132 | ||
BU707674.1 | - | 7 - 8 | 555 | 132 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protein Kinase Mammalian Gene Collection Mus musculus Probe spans exons |