Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prkag2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001170555.1 | NP_001164026.1 | 2 - 3 | 291 | 85 | |
NM_001170556.1 | NP_001164027.1 | 2 - 3 | 288 | 85 | ||
NM_145401.2 | NP_663376.2 | 6 - 7 | 1370 | 85 | ||
GenBank mRNA | AK010703.1 | - | 2 - 3 | 230 | 85 | |
AK032238.1 | - | 4 - 5 | 882 | 85 | ||
AK154296.1 | - | 6 - 7 | 1371 | 85 | ||
AK165834.1 | - | 2 - 3 | 230 | 85 | ||
AY348864.1 | - | 2 - 3 | 221 | 85 | ||
BC015283.1 | - | 6 - 7 | 1158 | 4238045 | 85 | |
BQ572978.1 | - | 2 - 3 | 221 | 85 | ||
BU707674.1 | - | 2 - 3 | 191 | 85 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protein Kinase Mammalian Gene Collection Mus musculus Probe spans exons |