Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Agap3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_139153.2 | NP_631892.2 | 12 - 13 | 1699 | 58 | |
GenBank mRNA | AF459091.1 | - | 12 - 13 | 1699 | 58 | |
AK079516.1 | - | 8 - 9 | 1038 | 58 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mus musculus Probe spans exons |