Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptpn12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011203.2 | NP_035333.2 | 15 - 16 | 2185 | 127 | |
GenBank mRNA | AK154107.1 | - | 15 - 16 | 2277 | 127 | |
BC051980.1 | - | 15 - 16 | 2185 | 6402545 | 127 | |
X63440.1 | - | 15 - 16 | 2180 | 127 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protein Phosphatase Mammalian Gene Collection Mus musculus Probe spans exons |