Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc16a7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011391.1 | NP_035521.1 | 5 - 6 | 796 | 76 | |
GenBank mRNA | AF058054.1 | - | 5 - 6 | 796 | 76 | |
AK076612.1 | - | 4 - 5 | 618 | 76 | ||
AK085398.1 | - | 4 - 5 | 76 | |||
BC089606.1 | - | 4 - 5 | 564 | 76 | ||
BC117803.1 | - | 4 - 5 | 560 | 40092297 | 76 | |
BC117804.1 | - | 4 - 5 | 560 | 40092299 | 76 | |
BC126960.1 | - | 4 - 5 | 560 | 40092496 | 76 | |
CT010191.1 | - | 2 - 3 | 359 | 76 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Transporter Mammalian Gene Collection Mus musculus Probe spans exons |