Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nrf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164226.1 | NP_001157698.1 | 9 - 10 | 1353 | 80 | |
NM_001164227.1 | NP_001157699.1 | 9 - 10 | 1353 | 80 | ||
NM_001164228.1 | NP_001157700.1 | 9 - 10 | 1350 | 80 | ||
NM_001164229.1 | NP_001157701.1 | 10 - 11 | 1427 | 80 | ||
NM_001164230.1 | NP_001157702.1 | 9 - 10 | 1350 | 80 | ||
NM_010938.4 | NP_035068.3 | 9 - 10 | 1350 | 80 | ||
GenBank mRNA | AF098077.1 | - | 9 - 10 | 1285 | 80 | |
AK014494.1 | - | 9 - 10 | 1314 | 80 | ||
AK029034.1 | - | 9 - 10 | 1331 | 80 | ||
AK031103.1 | - | 4 - 5 | 554 | 80 | ||
AK037697.1 | - | 9 - 10 | 1311 | 80 | ||
AK050861.1 | - | 9 - 10 | 1267 | 80 | ||
AK082580.1 | - | 9 - 10 | 1351 | 80 | ||
AK134483.1 | - | 9 - 10 | 1267 | 80 | ||
AK135673.1 | - | 10 - 11 | 1430 | 80 | ||
BC005410.1 | - | 8 - 9 | 1171 | 3496661 | 80 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |