Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zkscan3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001145778.1 | NP_001139250.1 | 4 - 5 | 776 | 113 | |
NM_023685.4 | NP_076174.3 | 4 - 5 | 773 | 113 | ||
GenBank mRNA | AK028902.1 | - | 4 - 5 | 763 | 113 | |
AK031080.1 | - | 4 - 5 | 734 | 113 | ||
AK031286.1 | - | 4 - 5 | 731 | 113 | ||
AK050028.1 | - | 4 - 5 | 735 | 113 | ||
AK182625.1 | - | 1 - 2 | 71 | 113 | ||
AK220177.1 | - | 4 - 5 | 940 | 113 | ||
BC007473.1 | - | 4 - 5 | 731 | 3496252 | 113 | |
BG296820.1 | - | 3 - 4 | 269 | 113 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |