Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpld1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008156.2 | NP_032182.2 | 24 - 25 | 2709 | 64 | |
GenBank mRNA | AK004957.2 | - | 23 - 24 | 2413 | 64 | |
AY081194.1 | - | 23 - 24 | 2436 | 64 | ||
AY333960.1 | - | 23 - 24 | 2393 | 64 | ||
BC019146.1 | - | 23 - 24 | 2422 | 5052822 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |