Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gdpd5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_201352.2 | NP_958740.2 | 7 - 8 | 1264 | 91 | |
GenBank mRNA | AK141189.1 | - | 7 - 8 | 1297 | 91 | |
AK154771.1 | - | 7 - 8 | 1401 | 91 | ||
BC024955.1 | - | 6 - 7 | 550 | 4953174 | 91 | |
BC026428.1 | - | 3 - 4 | 197 | 4224574 | 91 | |
BC082585.1 | - | 7 - 8 | 1264 | 30608279 | 91 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |