Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pgm2l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027629.3 | NP_081905.1 | 11 - 12 | 1758 | 154 | |
GenBank mRNA | AK016437.1 | - | 9 - 10 | 1117 | 154 | |
AK039189.1 | - | 11 - 12 | 1742 | 154 | ||
AK040513.1 | - | 10 - 11 | 1488 | 154 | ||
BC020351.1 | - | 11 - 12 | 1971 | 154 | ||
BC032150.1 | - | 7 - 8 | 964 | 5004308 | 154 | |
BC076571.1 | - | 11 - 12 | 1655 | 30356163 | 154 | |
BG071667.2 | - | 3 - 4 | 154 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |