Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pgm2l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027629.3 | NP_081905.1 | 1 - 2 | 426 | 145 | |
GenBank mRNA | AK039189.1 | - | 1 - 2 | 410 | 145 | |
AK040513.1 | - | 1 - 2 | 346 | 145 | ||
BB633030.1 | - | 1 - 2 | 407 | 145 | ||
BC020351.1 | - | 1 - 2 | 639 | 145 | ||
BC076571.1 | - | 1 - 2 | 323 | 30356163 | 145 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |