Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spcs2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025668.3 | NP_079944.1 | 3 - 4 | 359 | 110 | |
GenBank mRNA | AK014310.1 | - | 3 - 4 | 373 | 110 | |
AK017504.1 | - | 3 - 4 | 364 | 110 | ||
AK077510.1 | - | 3 - 4 | 369 | 110 | ||
AK134578.1 | - | 3 - 4 | 360 | 110 | ||
AK168173.1 | - | 3 - 4 | 364 | 110 | ||
BC010547.1 | - | 2 - 3 | 177 | 110 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protease Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |