Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chrdl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001291320.1 | NP_001278249.1 | 3 - 4 | 406 | 86 | |
NM_133709.3 | NP_598470.3 | 3 - 4 | 448 | 86 | ||
GenBank mRNA | AF338222.1 | - | 3 - 4 | 352 | 86 | |
BC019399.1 | - | 3 - 4 | 413 | 4012143 | 86 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |