Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trim3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001285870.1 | NP_001272799.1 | 5 - 6 | 928 | 68 | |
NM_001285871.1 | NP_001272800.1 | 5 - 6 | 1113 | 68 | ||
NM_001285873.1 | NP_001272802.1 | 5 - 6 | 928 | 68 | ||
NM_018880.3 | NP_061368.1 | 5 - 6 | 902 | 68 | ||
GenBank mRNA | AB030912.1 | - | 5 - 6 | 929 | 68 | |
AF220019.1 | - | 5 - 6 | 821 | 68 | ||
AK019165.1 | - | 5 - 6 | 863 | 68 | ||
AK144674.1 | - | 5 - 6 | 831 | 68 | ||
AK170029.1 | - | 5 - 6 | 866 | 68 | ||
BC003347.1 | - | 5 - 6 | 943 | 68 | ||
BC034263.1 | - | 5 - 6 | 867 | 4503884 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |