Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppfibp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163557.1 | NP_001157029.1 | 12 - 13 | 1257 | 65 | |
NM_008905.2 | NP_032931.2 | 12 - 13 | 1493 | 65 | ||
GenBank mRNA | AK012152.1 | - | 9 - 10 | 916 | 65 | |
BC055935.1 | - | 2 - 3 | 127 | 65 | ||
BC100346.1 | - | 12 - 13 | 1257 | 30936010 | 65 | |
U79024.1 | - | 12 - 13 | 1278 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |