Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cpa2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024698.2 | NP_001019869.1 | 4 - 5 | 396 | 75 | |
GenBank mRNA | BC094929.1 | - | 4 - 5 | 426 | 6434004 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protease Mammalian Gene Collection Mus musculus Probe spans exons |