Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mtdh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026002.4 | NP_080278.3 | 2 - 3 | 792 | 82 | |
GenBank mRNA | AF501309.1 | - | 2 - 3 | 730 | 82 | |
AK011808.1 | - | 2 - 3 | 787 | 82 | ||
AK029915.1 | - | 2 - 3 | 793 | 82 | ||
AK151269.1 | - | 2 - 3 | 776 | 82 | ||
AK151810.1 | - | 2 - 3 | 787 | 82 | ||
AY553638.1 | - | 2 - 3 | 475 | 82 | ||
BC023115.1 | - | 2 - 3 | 691 | 82 | ||
BC036994.1 | - | 2 - 3 | 511 | 82 | ||
BC138858.1 | - | 2 - 3 | 506 | 8861880 | 82 | |
BC138859.1 | - | 2 - 3 | 506 | 8861881 | 82 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |