Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pop1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026340.3 | NP_080616.1 | 10 - 11 | 1580 | 65 | |
NM_152894.2 | NP_690854.1 | 10 - 11 | 1580 | 65 | ||
GenBank mRNA | AK016547.1 | - | 10 - 11 | 1567 | 65 | |
BC034742.1 | - | 10 - 11 | 1559 | 4935194 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |