Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lat | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010689.2 | NP_034819.1 | 1 - 2 | 394 | 90 | |
GenBank mRNA | AF036907.1 | - | 1 - 2 | 394 | 90 | |
AK088763.1 | - | 1 - 2 | 303 | 90 | ||
AK134003.1 | - | 1 - 2 | 346 | 90 | ||
BC013337.1 | - | 1 - 2 | 371 | 4191983 | 90 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |