Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptpre | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011212.3 | NP_035342.3 | 5 - 6 | 606 | 149 | |
GenBank mRNA | AK004602.1 | - | 5 - 6 | 606 | 149 | |
AK132455.1 | - | 4 - 5 | 569 | 149 | ||
AK154910.1 | - | 2 - 3 | 158 | 149 | ||
AK160947.1 | - | 2 - 3 | 155 | 149 | ||
BC024631.1 | - | 5 - 6 | 600 | 149 | ||
BC053408.1 | - | 5 - 6 | 382 | 149 | ||
BC098473.1 | - | 5 - 6 | 370 | 149 | ||
D83484.1 | - | 5 - 6 | 943 | 149 | ||
U35368.1 | - | 5 - 6 | 606 | 149 | ||
U36758.1 | - | 2 - 3 | 155 | 149 | ||
U40280.1 | - | 4 - 5 | 346 | 149 | ||
U62387.1 | - | 5 - 6 | 508 | 149 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protein Phosphatase Mammalian Gene Collection Mus musculus Probe spans exons |