Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tubgcp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001286007.1 | NP_001272936.1 | 12 - 13 | 2263 | 66 | |
NM_001286009.1 | NP_001272938.1 | 12 - 13 | 2263 | 66 | ||
NM_001286011.1 | NP_001272940.1 | 11 - 12 | 2030 | 66 | ||
NM_133755.2 | NP_598516.2 | 12 - 13 | 2190 | 66 | ||
GenBank mRNA | AK006233.1 | - | 2 - 3 | 294 | 66 | |
AK033157.1 | - | 12 - 13 | 1980 | 66 | ||
AK045084.1 | - | 12 - 13 | 1979 | 66 | ||
AK146771.1 | - | 12 - 13 | 2185 | 66 | ||
AK158060.1 | - | 12 - 13 | 1979 | 66 | ||
AK167924.1 | - | 12 - 13 | 2085 | 66 | ||
BC012519.1 | - | 12 - 13 | 2118 | 3601589 | 66 | |
BC025564.1 | - | 11 - 12 | 1952 | 66 | ||
BC025582.1 | - | 12 - 13 | 2079 | 5324981 | 66 | |
BC027034.1 | - | 11 - 12 | 2464 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |