Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tspan33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001301407.1 | NP_001288336.1 | 6 - 7 | 821 | 72 | |
NM_146173.3 | NP_666285.1 | 6 - 7 | 824 | 72 | ||
GenBank mRNA | AF276890.1 | - | 6 - 7 | 823 | 72 | |
AK004954.1 | - | 6 - 7 | 797 | 72 | ||
AK172176.1 | - | 6 - 7 | 802 | 72 | ||
BC024685.1 | - | 6 - 7 | 782 | 3981492 | 72 | |
BE861472.1 | - | 4 - 5 | 433 | 72 | ||
BU054333.1 | - | 6 - 7 | 575 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |