Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atxn2l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_183020.1 | NP_898841.1 | 14 - 15 | 2081 | 87 | |
GenBank mRNA | AK155062.1 | - | 14 - 15 | 2128 | 87 | |
AK158626.1 | - | 14 - 15 | 1768 | 87 | ||
AK168745.1 | - | 14 - 15 | 1798 | 87 | ||
BC034083.1 | - | 8 - 9 | 1160 | 87 | ||
BC043451.1 | - | 9 - 10 | 1239 | 87 | ||
BC054483.1 | - | 14 - 15 | 2081 | 6391591 | 87 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |