Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gga2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028758.2 | NP_083034.1 | 15 - 16 | 1703 | 92 | |
GenBank mRNA | AK004632.1 | - | 15 - 16 | 1675 | 92 | |
AK034612.1 | - | 12 - 13 | 1346 | 92 | ||
AK129280.1 | - | 15 - 16 | 1662 | 92 | ||
AK143691.1 | - | 15 - 16 | 1691 | 92 | ||
AK149798.1 | - | 15 - 16 | 1693 | 92 | ||
AK154904.1 | - | 15 - 16 | 1678 | 92 | ||
AK159953.1 | - | 15 - 16 | 1693 | 92 | ||
AK211688.1 | - | 1 - 2 | 89 | 92 | ||
BC057377.1 | - | 8 - 9 | 1206 | 5698559 | 92 | |
BC062933.1 | - | 15 - 16 | 1672 | 30356060 | 92 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |