Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Inpp5f | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178641.5 | NP_848756.2 | 1 - 2 | 333 | 141 | |
GenBank mRNA | AK030870.1 | - | 1 - 2 | 335 | 141 | |
AK129249.1 | - | 1 - 2 | 290 | 141 | ||
AK154996.1 | - | 1 - 2 | 338 | 141 | ||
BC125437.1 | - | 1 - 2 | 221 | 40129952 | 141 | |
BC137700.1 | - | 1 - 2 | 221 | 8860719 | 141 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |