Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Srebf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011480.3 | NP_035610.1 | 7 - 8 | 1433 | 67 | |
GenBank mRNA | AF374266.1 | - | 6 - 7 | 1197 | 67 | |
AK052628.1 | - | 7 - 8 | 1434 | 67 | ||
AK150052.1 | - | 7 - 8 | 1422 | 67 | ||
AK154424.1 | - | 7 - 8 | 1401 | 67 | ||
AK169607.1 | - | 7 - 8 | 1434 | 67 | ||
BC006051.1 | - | 4 - 5 | 569 | 67 | ||
BC056922.1 | - | 7 - 8 | 1415 | 6824948 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |