Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tom1l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039092.3 | NP_001034181.1 | 9 - 10 | 1052 | 73 | |
NM_001039093.1 | NP_001034182.1 | 9 - 10 | 1052 | 73 | ||
NM_153080.2 | NP_694720.2 | 9 - 10 | 1052 | 73 | ||
GenBank mRNA | AF467887.1 | - | 9 - 10 | 1032 | 73 | |
AK043095.1 | - | 9 - 10 | 1032 | 73 | ||
AK054531.1 | - | 9 - 10 | 1020 | 73 | ||
AK166040.1 | - | 9 - 10 | 1052 | 73 | ||
BC062947.1 | - | 9 - 10 | 1016 | 30536491 | 73 | |
BC127266.1 | - | 9 - 10 | 1018 | 30357295 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Transporter Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |