Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ankhd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175375.3 | NP_780584.2 | 7 - 8 | 1387 | 60 | |
GenBank mRNA | AK131166.1 | - | 7 - 8 | 1250 | 60 | |
CF742822.1 | - | 2 - 3 | 259 | 60 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mus musculus Probe spans exons |