Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mvp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_080638.3 | NP_542369.2 | 4 - 5 | 548 | 64 | |
GenBank mRNA | AF210456.1 | - | 4 - 5 | 550 | 64 | |
AK088046.1 | - | 4 - 5 | 533 | 64 | ||
AK152534.1 | - | 4 - 5 | 523 | 64 | ||
AK167899.1 | - | 4 - 5 | 524 | 64 | ||
AK168100.1 | - | 4 - 5 | 524 | 64 | ||
BC006709.1 | - | 4 - 5 | 513 | 3584439 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |