Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dpagt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007875.2 | NP_031901.2 | 7 - 8 | 1204 | 128 | |
GenBank mRNA | AK008246.1 | - | 6 - 7 | 1056 | 128 | |
AK008317.1 | - | 6 - 7 | 1057 | 128 | ||
BC003943.1 | - | 7 - 8 | 861 | 3487188 | 128 | |
BC010474.1 | - | 7 - 8 | 1204 | 3965178 | 128 | |
X65603.1 | - | 7 - 8 | 1069 | 128 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |