Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vps11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027889.1 | NP_082165.1 | 12 - 13 | 2101 | 90 | |
GenBank mRNA | AK004695.1 | - | 12 - 13 | 2076 | 90 | |
AK032484.1 | - | 12 - 13 | 2066 | 90 | ||
AK037658.1 | - | 10 - 11 | 2556 | 90 | ||
AK090030.1 | - | 12 - 13 | 2082 | 90 | ||
AK153963.1 | - | 12 - 13 | 2074 | 90 | ||
AK162897.1 | - | 12 - 13 | 2076 | 90 | ||
AK207095.1 | - | 1 - 2 | 80 | 90 | ||
BC029004.1 | - | 12 - 13 | 2101 | 5360111 | 90 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |