Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0900017966.mm.1 |
NM_027889.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900017966.mm.1 |
NM_027889.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900017966.mm.1 |
TC0900002159.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900017959.mm.1 |
NM_027889.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900017959.mm.1 |
NM_027889.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900017959.mm.1 |
TC0900002159.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900017964.mm.1 |
NM_027889.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900017964.mm.1 |
NM_027889.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900017964.mm.1 |
TC0900002159.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900017960.mm.1 |
NM_027889.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900017960.mm.1 |
NM_027889.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900017960.mm.1 |
TC0900002159.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900017961.mm.1 |
NM_027889.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900017961.mm.1 |
NM_027889.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900017961.mm.1 |
TC0900002159.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900017965.mm.1 |
NM_027889.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900017965.mm.1 |
NM_027889.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900017965.mm.1 |
TC0900002159.mm.1 |
MTA TC |