Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Polrmt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172551.3 | NP_766139.2 | 8 - 9 | 1571 | 65 | |
GenBank mRNA | AK042071.1 | - | 6 - 7 | 1128 | 65 | |
AK047545.1 | - | 8 - 9 | 1564 | 65 | ||
AK053356.1 | - | 8 - 9 | 1562 | 65 | ||
AK085666.1 | - | 8 - 9 | 1564 | 65 | ||
AK128956.1 | - | 8 - 9 | 1563 | 65 | ||
AK154181.1 | - | 8 - 9 | 1562 | 65 | ||
AK154729.1 | - | 8 - 9 | 1553 | 65 | ||
AK213762.1 | - | 1 - 2 | 96 | 65 | ||
BC110697.1 | - | 8 - 9 | 1547 | 30356222 | 65 | |
BC117939.1 | - | 8 - 9 | 1564 | 65 | ||
BC117940.1 | - | 8 - 9 | 1564 | 40097816 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |