Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vps26a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113355.1 | NP_001106826.1 | 2 - 3 | 371 | 136 | |
NM_133672.3 | NP_598433.1 | 2 - 3 | 298 | 136 | ||
GenBank mRNA | AK028096.1 | - | 2 - 3 | 372 | 136 | |
AK086851.1 | - | 194 | 136 | |||
AK147989.1 | - | 2 - 3 | 239 | 136 | ||
AK149859.1 | - | 2 - 3 | 223 | 136 | ||
AK150244.1 | - | 2 - 3 | 196 | 136 | ||
AK151360.1 | - | 2 - 3 | 208 | 136 | ||
AK152424.1 | - | 2 - 3 | 242 | 136 | ||
AK159783.1 | - | 2 - 3 | 248 | 136 | ||
AK165853.1 | - | 2 - 3 | 242 | 136 | ||
AK167592.1 | - | 2 - 3 | 253 | 136 | ||
AK168213.1 | - | 2 - 3 | 298 | 136 | ||
AK168537.1 | - | 2 - 3 | 253 | 136 | ||
AK209699.1 | - | 1 - 2 | 161 | 136 | ||
AK216993.1 | - | 1 - 2 | 161 | 136 | ||
BB857335.1 | - | 2 - 3 | 136 | |||
BC007148.1 | - | 2 - 3 | 219 | 3485844 | 136 | |
S41204.1 | - | 2 - 3 | 245 | 136 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |