Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccar1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026201.3 | NP_080477.1 | 14 - 15 | 2020 | 97 | |
GenBank mRNA | AF465615.1 | - | 14 - 15 | 1911 | 97 | |
AK012111.1 | - | 14 - 15 | 1911 | 97 | ||
BC010199.1 | - | 14 - 15 | 1898 | 97 | ||
BC034174.1 | - | 14 - 15 | 1921 | 97 | ||
BC039939.1 | - | 14 - 15 | 1921 | 97 | ||
BC060130.1 | - | 14 - 15 | 1908 | 97 | ||
BC079652.1 | - | 14 - 15 | 1894 | 6844310 | 97 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |