Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snx14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172926.3 | NP_766514.2 | 4 - 5 | 562 | 94 | |
GenBank mRNA | AK046839.1 | - | 4 - 5 | 504 | 94 | |
AK049371.1 | - | 4 - 5 | 541 | 94 | ||
AK082389.1 | - | 4 - 5 | 541 | 94 | ||
BC080799.1 | - | 4 - 5 | 547 | 4948538 | 94 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |