Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snap91 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277982.1 | NP_001264911.1 | 7 - 8 | 763 | 132 | |
NM_001277983.1 | NP_001264912.1 | 7 - 8 | 875 | 132 | ||
NM_001277985.1 | NP_001264914.1 | 7 - 8 | 875 | 132 | ||
NM_001277986.1 | NP_001264915.1 | 7 - 8 | 875 | 132 | ||
NM_013669.2 | NP_038697.1 | 7 - 8 | 875 | 132 | ||
NR_102729.1 | 7 - 8 | 875 | 132 | |||
GenBank mRNA | AK051176.1 | - | 7 - 8 | 870 | 132 | |
AK147087.1 | - | 7 - 8 | 866 | 132 | ||
AK159570.1 | - | 7 - 8 | 841 | 132 | ||
BC031773.1 | - | 7 - 8 | 763 | 4511467 | 132 | |
BC052454.1 | - | 7 - 8 | 875 | 6410678 | 132 | |
M83985.1 | - | 7 - 8 | 800 | 132 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |