Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Piwil4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177905.3 | NP_808573.2 | 12 - 13 | 1766 | 100 | |
GenBank mRNA | AB258534.1 | - | 12 - 13 | 1724 | 100 | |
AK143022.1 | - | 3 - 4 | 536 | 101 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Amplicon length greater than or equal to 101 Mus musculus Probe spans exons |