Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Boc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172506.2 | NP_766094.1 | 8 - 9 | 2055 | 68 | |
GenBank mRNA | AF388037.1 | - | 7 - 8 | 1508 | 68 | |
AK028770.1 | - | 8 - 9 | 2076 | 68 | ||
AK028889.1 | - | 8 - 9 | 2056 | 68 | ||
BC062892.1 | - | 8 - 9 | 2035 | 6418202 | 68 | |
BC078631.1 | - | 8 - 9 | 1999 | 6515418 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |