Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Loxl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010729.3 | NP_034859.2 | 1 - 2 | 1526 | 74 | |
GenBank mRNA | AF357006.1 | - | 1 - 2 | 1287 | 74 | |
AK078512.1 | - | 1 - 2 | 1515 | 74 | ||
BC003973.1 | - | 1 - 2 | 581 | 74 | ||
BC037999.1 | - | 1 - 2 | 1510 | 5251968 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |