Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1000018673.mm.1 |
NM_145361.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000018673.mm.1 |
NM_145361.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000018673.mm.1 |
TC1000002508.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000018677.mm.1 |
NM_145361.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000018677.mm.1 |
NM_145361.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000018677.mm.1 |
TC1000002508.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000018676.mm.1 |
NM_145361.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000018676.mm.1 |
NM_145361.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000018676.mm.1 |
TC1000002508.mm.1 |
MTA TC |