Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atp8b3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026094.3 | NP_080370.2 | 7 - 8 | 608 | 93 | |
GenBank mRNA | AY364445.1 | - | 7 - 8 | 608 | 93 | |
BC118977.1 | - | 7 - 8 | 608 | 40098417 | 93 | |
BC118978.1 | - | 7 - 8 | 608 | 40098419 | 93 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Transporter Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |