Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gnb5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010313.2 | NP_034443.1 | 5 - 6 | 64 | ||
NM_138719.5 | NP_619733.1 | 3 - 4 | 64 | |||
NR_073186.1 | 4 - 5 | 64 | ||||
GenBank mRNA | AK018747.1 | - | 1 - 2 | 64 | ||
AK050564.1 | - | 3 - 4 | 64 | |||
AK148175.1 | - | 3 - 4 | 393 | 64 | ||
AK149908.1 | - | 3 - 4 | 64 | |||
AK169323.1 | - | 3 - 4 | 64 | |||
BC016135.1 | - | 5 - 6 | 4506201 | 64 | ||
CK621143.1 | - | 5 - 6 | 64 | |||
L34290.1 | - | 3 - 4 | 64 | |||
U69145.1 | - | 4 - 5 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |