Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ubxn7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177633.4 | NP_808301.3 | 8 - 9 | 873 | 83 | |
GenBank mRNA | AK129218.1 | - | 4 - 5 | 471 | 83 | |
AK147730.1 | - | 8 - 9 | 869 | 83 | ||
BC062904.1 | - | 7 - 8 | 791 | 83 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Apoptosis Mammalian Gene Collection Mus musculus Probe spans exons |